Learn More
Bollé Safety Prism Smoke Safety Glasses
Click to view available options
Frame Material:
Polycarbonate
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Smoke |
Product Type | Prism PRIPSF Safety Spectacles |
No. per Pack | 1 Ea. |
Weight (Metric) | 23 g |
Frame Material | Polycarbonate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Documents
Product Information
Specification Sheets
PRISM
File size: 509 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Bollé Safety Prism Smoke Safety Glasses >
Spot an opportunity for improvement?Share a Content Correction