missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propantheline bromide, 99.72%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
100g
Specifications
Specifications
CAS | 50-34-0 |
Color | White to White-Yellow |
Molecular Formula | C23H30BrNO3 |
Quantity | 100 g |
Synonym | propantheline bromide, pro-banthine, neometantyl, neopepulsan, corrigast, ercorax, ercotina, ketaman, kivatin |
InChI Key | XLBIBBZXLMYSFF-UHFFFAOYSA-M |
SMILES | CC(C)[N+](C)(CCOC(=O)C1C2=CC=CC=C2OC3=CC=CC=C13)C(C)C.[Br-] |
IUPAC Name | methyl-di(propan-2-yl)-[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium;bromide |
Molecular Weight (g/mol) | 448.401 |
PubChem CID | 9279 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Propantheline bromide, 99.72%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction