Learn More
Invitrogen™ ProtoArray™ Blocking Buffer Kit
The Blocking Buffer Kit consists of reagents sufficient for preparing Blocking Buffer and Wash Buffer to process 10 ProtoArray™ Human Protein Microarrays.
Brand: Invitrogen™ PA055
Description
The Blocking Buffer Kit consists of reagents sufficient for preparing Blocking Buffer and Wash Buffer to process 10 ProtoArray™ Human Protein Microarrays for ProtoArray™ protein-protein interaction (PPI), immune response biomarker profiling (IRBP), and antibody specificity profiling (ASP) applications. The kit contains one bottle each of Blocking Buffer and 10X Synthetic Block, and one tube of dithiothreitol (DTT).
Specifications
ProtoArray™ Blocking Buffer Kit, 1 Kit, –20°C Storage Temperature, For Applications Pharma and Biopharma, Protein Assays and Analysis, Target And Lead Identification And Validation | |
Protein Array Blocking Buffer | |
Protein Assay and Analysis | |
ProtoArray™ Human Protein Microarrays for immune response biomarker profiling (IRBP), ProtoArray™ Human Protein Microarrays for antibody specificity profiling (ASP), ProtoArray™ Human Protein Microarrays for protein-protein interaction (PPI) |
Blocking Buffer Kit | |
1 Kit | |
Store the ProtoArray™ Blocking Buffer Kit at -20°C., -20°C | |
ProtoArray™ |