missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pseudomonas CFC Selective Medium

Selectively isolate Pseudomonas species from a range of samples with this ready-to-use medium.

Brand:  Thermo Scientific™ PO5132A

12.51 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 10474203

  • 13.75 EUR / Pack of 10
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Selectively isolate Pseudomonas species from a range of samples with ready-to-use Thermo Scientific™ Oxoid™ Pseudomonas CFC Selective Medium. The medium contains magnesium chloride and potassium sulphate for improved pigment production and recovery of Pseudomonas aeruginosa or Pseudomonas spp. while strongly suppressing Klebsiella, Proteus and Providencia spp.

Use Pseudomonas CFC Selective Agar for selective isolation of Pseudomonas species from a range of samples.

  • Enhanced pigment production aids identification
  • Pseudomonas aeruginos produce characteristic bright green colour
  • KlebsiellaProteus and Providencia are strongly suppressed
  • Ready-to-use convenience of prepared media

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Oyster white, transparent
Pseudomonas CFC Selective Medium
Germany
7.1 ± 0.2
10/Pk.
6°C to 12°C
90mm Plate
Agar
Flow wrapped - Single Layer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6252074 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberPO5132A
Lot Number6252073 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberPO5132A
Lot Number6249174 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPO5132A
Lot Number6245985 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberPO5132A
Lot Number6242478 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog NumberPO5132A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pseudomonas CFC Selective Medium >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.