Learn More
Fisherbrand™ PTFE Blade for Plain Shafts
Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals. Cowie Technology™ PTFE Blade for Plain Shafts fit all standard shafts for blades.
Brand:  Fisherbrand™ 002.1105.1
 View more versions of this product
Product Code. 16205861
  • 39.90 EUR / Pack of 10
Estimated Shipment: 23-06-2025
Description
  • Made of PTFE
  • Totally inert and non-scratch
  • Available in square end and plain end format
Specifications
Specifications
PTFE Shaft Stirrer, Plain Shaft for Blades
25 mm
-200°C to +280°C
Blades for Plain Shafts
Stirring in Narrow Neck Vessel
1 Each
3 mm
105 mm
Documents
One moment while we fetch your results.
Certificates
View accessory products
Product Title
Fisherbrand™ PTFE Blade for Plain Shafts > Width: 105mm