Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bars
Standard stir bars with an Alnico 5 magnetic core encapsulated in PTFE.
Brand: Bohlender™ C350-07
This item is not returnable.
View return policy
Description
- Providing temperature resistance from −200° to +250°C
- Universal chemical resistance
- FDA conformity
- Offers excellent centering and smooth running characteristics
- Available in a wide range of sizes
Specifications
FDA | |
Alnico 5 magnetic core | |
8 mm | |
PTFE |
White | |
−200° to +250°C | |
3 mm | |
Cylinder |