Learn More
Fisherbrand™ PTFE Plain Stirrer Shaft for Blades
Use with PTFE blades. The PTFE Plain Stirrer Shaft for Blades include a steel rod core.
28.75 EUR - 84.90 EUR
Specifications
For Use With (Equipment) | PTFE Stirrer Blades |
---|---|
For Use With (Application) | Mixing |
Quantity | 1 Each |
Temperature (Metric) | -200°C to +280°C |
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Shaft | Length (Metric) | Price | Quantity & Availability | |||||
10148181
|
Fisherbrand™
005.550.8.1 |
8 mm | 500 mm |
40.70 EUR
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
10239321
|
Fisherbrand™
005.555.1 |
9.5 mm | 550 mm |
37.60 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10168131
|
Fisherbrand™
005.530.1 |
6 mm | 300 mm |
28.75 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10720723
|
Fisherbrand™
005.540.1 |
6 mm | 400 mm |
29.30 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10138181
|
Fisherbrand™
005.540.8.1 |
8 mm | 400 mm |
39.95 EUR
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
|||||
10056031
|
Fisherbrand™
005.550.1 |
6 mm | 500 mm |
31.55 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10341352
|
Fisherbrand™
005.530.8.1 |
8 mm | 300 mm |
39.50 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10761293
|
Fisherbrand™
005.565.1 |
9.5 mm | 650 mm |
38.35 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
10065961
|
Fisherbrand™
005.0565.1 |
9.5 mm | 650 mm (EX) |
42.55 EUR
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
10085971
|
Fisherbrand™
005.0575.1 |
9.5 mm | 750 mm (EX) |
43.35 EUR
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
13154299
|
Fisherbrand™
005.05100.10.1 |
10 mm | 1000 mm (EX) |
68.15 EUR
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
|||||
13164299
|
Fisherbrand™
005.05100.12.1 |
12 mm | 1000 mm (EX) |
84.90 EUR
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
10595872
|
Fisherbrand™
005.0565.10.1 |
10 mm | 650 mm (EX) |
47.45 EUR
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
|||||
10187941
|
Fisherbrand™
005.0575.12.1 |
12 mm | 750 mm (EX) |
79.10 EUR
Each |
Estimated Shipment: 23-07-2025
Log in to see stock. |
|||||
13174299
|
Fisherbrand™
005.0565.12.1 |
12 mm | 650 mm (EX) |
82.10 EUR
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10759952
|
Fisherbrand™
005.0575.10.1 |
10 mm | 750 mm (EX) |
49.00 EUR
Each |
Estimated Shipment: 26-06-2025
Log in to see stock. |
|||||
Description
PTFE shafts have a fully PTFE-encapsulated steel reinforced drive shaft.- Chemically resistant, inert, and leak-free
- Easy to clean
- No cross contamination
- Shatterproof
- Non-scratch
- Use in temperatures up to approximately 280°C
- Exposed shaft models (EX) are produced with a stainless steel core with an exposed end for more rigid clamping to the chuck drive
Specifications
PTFE Stirrer Blades | |
Mixing | |
1 Each | |
-200°C to +280°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ PTFE Plain Stirrer Shaft for Blades