Learn More
Fisherbrand™ PTFE Stopper, Flat Head, Full Length A Type
Fisherbrand™ PTFE Stopper, Flat Head fit standard taper ground glass joints. Trapezoidal rings give high vacuum performance and reduce seizure.
Brand:  Fisherbrand™ 010.119
 View more versions of this product
Product Code. 16205951
  • 13.65 EUR / Each
Estimated Shipment: 11-11-2024
Description
  • Solid Construction
  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications
Specifications
Fisherbrand™ PTFE Stopper, Flat Head, Full Length A Type
50 mm
19/38
PTFE
Standard Taper Ground Glass Joints
1
-200°C to +280°C