Learn More
Julabo™ PURA 4 Water Bath
PURA water baths stand for a straightforward and safe working in the laboratory
Brand: Julabo™ 9550404
Description
The high-quality water baths offer a temperature stability of ± 0.15 °C. The working temperature range of +18 °C to 99.9 °C allows a wide range of applications, such as routine laboratory work, temperature applications for samples, incubation, material testing, corrosion testing, temperature control applications of cultures as well as temperature tests of food and beverages and in the dental field.
PURA water baths can be used flexibly and can be easily transported through ergonomic side recessed grips. Non-slip rubber feet ensure a firm footing and more safety in the laboratory.
The units are splash-proof, feature a (membrane) keypad and temperature display (LED) for ease of use. There are no disturbing parts inside the bath so that the entire inner surface can be used to safely temperature-control even small sample containers.
Advantages:
- Ease of use
- Bright display
- Splash-proof protected mains switch
- Built-in dry running protection
- No disturbing elements in the bath
- Platform included as standard
- Integrated timer
- Non-slip rubber feet
- Ergonomic recessed grips
- Space-saving model for use with one test tube rack
Specifications
Water Bath | |
LED Display | |
35 x 21 x 22 cm |
0.8 to 4.5 L | |
Acoustic and optical alarm signal |