Learn More
Moldex™ Pura-Fit MoldexStations
Click to view available options
Size: Large
Description
Made from recyclable material and with only 2% wastage the dispenser puts protection at the center of the workforce. Plugs remain hygienic as the dispenser mechanism is automatically replaced with every refill. Contains 250 pairs.
Specifications
Specifications
Includes | 250 Pairs of Moldex Pura-Fit earplugs |
Size | Large |
Quantity | 250/Pk. |
Noise Reduction Rating (NRR) | 36 dB |
Product Type | Pura-Fit MoldexStation |