Learn More
Deutsch Neumann™ Atomizer Bulbs
Excellent for producing fine sprays of liquid in the laboratory. Deutsch Neumann™ Atomizer Bulbs are manufactured from soft PVC.
Brand: Deutsch Neumann™ 4200003
7.44 EUR valid until 2025-03-14
Use promo code "24150" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reddish-brown color
- Pressure valve
Specifications
Red/Brown | |
1 Pack |
Product Content Correction
Product Title
Deutsch Neumann™ Atomizer Bulbs > Capacity: 65mL
Spot an opportunity for improvement?Share a Content Correction