Learn More
Hanna Instruments™ PVDF Body 7 Pin Electrode
Brand: Hanna Instruments™ FC202D
This item is not returnable.
View return policy
Specifications
DIN | |
Used for sensing pH, bulb shape, body material, type of junction, type of reference and electrolyte used are just some of the design considerations | |
0 to 12 |
7 Pin Electrode | |
PVDF Body | |
0°C to 50°C |
Product Content Correction
Product Title
Hanna Instruments™ PVDF Body 7 Pin Electrode > 7 Pin Electrode
Spot an opportunity for improvement?Share a Content Correction