Learn More
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper
Manufactured from chemically resistant Pyrex™ borosilicate glass with hexagonal glass.
170.00 EUR - 750.00 EUR
Specifications
Base Type | Hexagonal |
---|---|
Type | Graduated Cylinder |
Material | Glass |
Certifications/Compliance | ISO 4788 |
Class | A |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15707957
|
Pyrex
3212/12WC |
250 mL |
660.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15727947
|
Pyrex
3212/04WC |
10 mL |
170.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15727957
|
Pyrex
3212/14WC |
500 mL |
292.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15747947
|
Pyrex
3212/06WC |
25 mL |
221.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15747957
|
Pyrex
3212/16WC |
1000 mL |
339.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15767947
|
Pyrex
3212/08WC |
50 mL |
510.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15767957
|
Pyrex
3212/18WC |
2000 mL |
264.00 EUR
Each |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
15787947
|
Pyrex
3212/10WC |
100 mL |
750.00 EUR
Pack of 2 |
Estimated Shipment: 20-08-2025
Log in to see stock. |
||||||
Description
- Complies with ISO 4788
- Calibrated to Class A tolerances
- Individually tested and supplied with certificate showing exact volume contained at 5 points on the scale
- Conformity H certified
- Printed with individual serial number for batch certification
- Manufactured from chemically resistant Pyrex borosilicate glass with hexagonal glass
- Graduations in white enamel for improved visibility with dark colored solutions
- Supplied with polyethylene stopper
Specifications
Hexagonal | |
Glass | |
A | |
Polyethylene |
Graduated Cylinder | |
ISO 4788 | |
Polyethylene Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Pyrex™ Class A Work Certified Glass Cylinder with Polyethylene Stopper