Learn More
Fisherbrand™ Pyrex™ Magnetic Stir Bar
Useful for high temperature applications up to 274°C. Fisherbrand™ Pyrex™ Magnetic Stir Bars have a completely smooth surface offering zero absorption and are easy to clean.
Brand: Fisherbrand™ 371015008
21.23 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Autoclavable
- Glass encapsulated Alnico magnet
- Manufactured in registered ISO 9001:2008 facilities
- Meets US Pharmacopoeia specifications for Type I borosilicate glass
Specifications
Autoclavable |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%