Learn More
Pyridine-N-oxide, 95%, Thermo Scientific Chemicals
37.00 EUR - 259.29 EUR
Chemical Identifiers
CAS | 694-59-7 |
---|---|
Molecular Formula | C5H5NO |
Molecular Weight (g/mol) | 95.1 |
MDL Number | MFCD00006194 |
InChI Key | ILVXOBCQQYKLDS-UHFFFAOYSA-N |
Synonym | pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide |
PubChem CID | 12753 |
ChEBI | CHEBI:29136 |
IUPAC Name | 1-oxidopyridin-1-ium |
SMILES | C1=CC=[N+](C=C1)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10331184
|
Thermo Scientific Acros
157690250 |
25 g | Glass bottle |
37.00 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645402
|
Thermo Scientific Acros
157691000 |
100 g | Glass bottle |
78.79 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001830
|
Thermo Scientific Acros
157695000 |
500 g | Glass bottle |
259.29 EUR
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
694-59-7 | |
95.1 | |
ILVXOBCQQYKLDS-UHFFFAOYSA-N | |
12753 | |
1-oxidopyridin-1-ium |
C5H5NO | |
MFCD00006194 | |
pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide | |
CHEBI:29136 | |
C1=CC=[N+](C=C1)[O-] |
Specifications
694-59-7 | |
Brown to White | |
143°C | |
95% | |
MFCD00006194 | |
01,966; 02,353; 05,567; 09,396 | |
pyridine-n-oxide, pyridine 1-oxide, pyridine n-oxide, pyridine oxide, pyridine, 1-oxide, pyridine-1-oxide, unii-91f12jjj4h, py n-oxide, pyridin-1-ium-1-olate, pyridine-oxide | |
ILVXOBCQQYKLDS-UHFFFAOYSA-N | |
1-oxidopyridin-1-ium | |
12753 | |
95.1 | |
Crystalline Solid, Crystals, or Chunks |
60.0°C to 65.0°C | |
270.0°C | |
Authentic | |
C5H5NO | |
20, II, 131 | |
15, 8084 | |
Solubility in water: soluble. | |
C1=CC=[N+](C=C1)[O-] | |
95.1 | |
CHEBI:29136 | |
95% | |
Pyridine-N-oxide, 98% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
missing translation for 'einecsNumber' : 211-774-6
missing translation for 'rtecsNumber' : UT6410000
missing translation for 'tsca' : TSCA
- Pyridine-N-oxide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.