missing translation for 'onlineSavingsMsg'
Learn More

Spectroline™ Q-Series UV Magnifier Lamp

Designed to detect trace amounts of fluorescence through a built-in 3X magnifier lens

Brand:  Spectroline™ Q-22B

Encompass

 View more versions of this product

Product Code. 11586696

  • POA / Each

This item requires a price update prior to being ordered.

Request a Price for this item here.

View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Longwave (LW) tubes are useful in medical diagnostic applications. Applications for the shortwave (SW) tubes include chromatography and UV sanitization. White light is for all types of visual inspection. All models have a high-impact polycarbonate housing that encloses a specular aluminum reflector to ensure maximum irradiance.

†Relative intensity at 6 in. (15cm).

Order Info

Overall dimensions: 7L x 5 3/4W x 2 1/4 in.H (17.8 x 14.6 x 5.7cm).

Specifications

Specifications

Polycarbonate
2.25 in.
Four 4 W longwave UV-A tubes
177.8 mm
5.75 in.
7 x 5.75 x 2.25 in. (177.8 x 146.05 x 57.15 mm)
57.15 mm
7 in.
1 Ea.
146.05 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.