Learn More
Socorex™ Qualitips Autoclaving Boxes
Brand:  Socorex™ 312.10ER
Product Code. 11771248
  • 65.40 EUR / Pack of 4
Estimated Shipment: 11-06-2025
Specifications
Macro Tip 10 mL
4 Pk.
Product Title
Socorex™ Qualitips Autoclaving Boxes > For 10mL Tip; Quantity: 4 Pack