Learn More
Munktell Filtrak™ Quantitative Ashless Papers
Recommended for use where a high purity is required for quantitative analysis. Munktell Filtrak™ Quantitative Ashless Papers; Circle are acid-washed to remove impurities and are made using pure cellulose with an alpha-cellulose content of almost 100%.
Brand:  Munktell Filtrak™ 100052
 View more versions of this product
Product Code. 11302994
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
  • Very fast filtering, open fiber matrix
  • Fast filtering, open to semi-open fiber matrix
  • Medium to fast filtering, semi-open to open fiber matrix
  • Medium to slow filtering, medium-tight fiber matrix
  • Very slow filtration, very tight fiber matrix
Specifications
Specifications
Medium Crystalline
8 to 10 μm
Medium to fast filtering, medium wide pores, typical ashless grade for quantitative tasks, coarser precipitates such as lead
240 mm
Cellulose
13 sec./10 mL
OOM