Learn More
Munktell Filtrak™ Quantitative Ashless Papers
Recommended for use where a high purity is required for quantitative analysis. Munktell Filtrak™ Quantitative Ashless Papers; Circle are acid-washed to remove impurities and are made using pure cellulose with an alpha-cellulose content of almost 100%.
Brand: Munktell Filtrak™ 100052
This item is not returnable.
View return policy
Description
- Very fast filtering, open fiber matrix
- Fast filtering, open to semi-open fiber matrix
- Medium to fast filtering, semi-open to open fiber matrix
- Medium to slow filtering, medium-tight fiber matrix
- Very slow filtration, very tight fiber matrix
Specifications
Medium Crystalline | |
8 to 10 μm | |
Medium to fast filtering, medium wide pores, typical ashless grade for quantitative tasks, coarser precipitates such as lead | |
240 mm |
Cellulose | |
13 sec./10 mL | |
OOM |