Learn More
Munktell Filtrak™ Quantitative Ashless Papers
Recommended for use where a high purity is required for quantitative analysis. Munktell Filtrak™ Quantitative Ashless Papers; Circle are acid-washed to remove impurities and are made using pure cellulose with an alpha-cellulose content of almost 100%.
Brand: Munktell Filtrak™ 100060
This item is not returnable.
View return policy
Description
- Very fast filtering, open fiber matrix
- Fast filtering, open to semi-open fiber matrix
- Medium to fast filtering, semi-open to open fiber matrix
- Medium to slow filtering, medium-tight fiber matrix
- Very slow filtration, very tight fiber matrix
Specifications
Coarse Crystalline | |
>10 μm | |
For coarse and voluminous precipitates such as iron, aluminum, and chromium hydroxide; Si-determination in steel and pig iron analysis | |
150 mm |
Cellulose | |
11s/10mL | |
OOR |