Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Tac System Extended Warranty Package
Adds innovative smart features to the real-time PCR (qPCR) workflow, all in a compact footprint.
Brand: Applied Biosystems™ A47651
This item is not returnable.
View return policy
Description
- Includes facial authentication, voice commands, Smart Help, multiplex capability, motorized interchangeable blocks, simplified workflow, intuitive software, and large capacitive touch-screen interface. Provides exceptional reproducibility with minimal well-to-well and instrument-to-instrument variation. Designed for ultimate ease-of-use, the system offers high quality, excellent reliability, and an optimal user experience for researchers who want to work smarter in the lab.
- Easy and convenient hands-free operation with facial authentication and voice commands
- Reduced errors and manual entries with smart qPCR workflow and TaqMan Array Plates with RFID
- Space savings with small footprint and true standalone operation; set-up, start, monitor, and analyze directly on the touchscreen
- Application versatility with easily changed blocks and four different block types
- Increased productivity with automation compatibility for 24/7 operation
- Access, analyze, and share data anytime, anywhere with our Connect cloud-based platform for real-time information access, secure data storage, and online results sharing
- Remotely monitor experiments using the Instrument Connect monitoring app
- Design and Analysis software compatible with PC and Mac operating systems
- Compatible with over 8 million TaqMan Assays for gene expression, genetic variation, gene regulation, or protein expression experiments
- The Connect platform provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the platform are web browser-based software tools that are flexible, fast, and easy to use. These software tools support a variety of analyses including relative quantitation, standard curve, high resolution melt (HRM), genotyping, and presence/absence. They are compatible with Mac or PC as well as most Applied Biosystems instrument file types.
- The system is delivered with one block type; other block types may be purchased separately. In addition, an Orbitor Microplate Mover is available for this system to support high-throughput workflows. For a more value-priced system that does not support TaqMan array cards or automation (but that can be upgraded to do so at a later date), please see our QuantStudio 6 Pro Real-Time PCR System. And for the ultimate in flexibility, consider the QuantStudio 12K Flex Real-Time PCR System, which also accommodates TaqMan OpenArray plates.
- Run hundreds of real-time PCR reactions effortlessly using TaqMan Array 384-well microfluidic cards. And when integrated with the Orbitor Microplate Mover (available separately), the QuantStudio 7 Pro system maximizes your throughput capabilities.
- Simple workflows, reliable assays and reagents, multiple formats, and intuitive software are available for a broad range of applications, including gene expression, genetic variation, and gene regulation.
- Improved well-to-well and instrument-to-instrument data accuracy is achieved with the OptiFlex system, which features six decoupled excitation and emission filter channels with 21 filter combinations for maximum multiplexing and chemistry flexibility.
- Detect changes in target quantity as small as 1.5-fold in single-plex reactions and obtain 10 logs of linear dynamic range.
Specifications
Interchangeable | |
QuantStudio™ | |
Room Temperature |
QuantStudio™, ViiA™ 7 System | |
Touchscreen |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio™ 7 Pro Tac System Extended Warranty Package > QuantStudio™, ViiA™ 7 System
Spot an opportunity for improvement?Share a Content Correction