Learn More
Invitrogen™ Qubit™ Flex Pyrogen-Free Tube Strips
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps.
Brand:  Invitrogen™ Q32893
203.68 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 17825247
  • 281.00 EUR / Pack of 120
In Stock
Description
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and the Qubit Endotoxin Detection Assay Kit (Cat. No. Q32891).
Notes
• For DNA, RNA, and protein quantification use Qubit Flex Assay Tube Strips (Cat. No. Q33252).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Assay Tube Strip
120 strips
Qubit Flex Fluorometer
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3124393 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberQ32893
Lot Number3097671 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberQ32893
Lot Number3097670 Certificate TypeCertificate of Analysis Date29/12/2024 Catalog NumberQ32893
Lot Number3097669 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberQ32893
Lot Number3097667 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog NumberQ32893
Product Title
Invitrogen™ Qubit™ Flex Pyrogen-Free Tube Strips > 120 Tube Strips