Learn More
Quickfit™ Borosilicate Glass Dufton Fractionating Column
Efficient fractionating column designed with an an accurately fitted Dufton glass spiral. Quickfit™ Borosilicate Glass Dufton Fractionating Column can be disassembled for cleaning.
Brand: Quickfit™ FC1/12
131.10 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Glass column with accurately fitted Dufton glass spiral
- Good efficiency - liquid spiralling down column meets ascending vapour
- Spiral can be removed for cleaning
Specifications
270mm | |
Dufton Fractionating Column | |
Borosilicate Glass | |
ISO 3585 |