Learn More
Quickfit™ Borosilicate Glass Dufton Fractionating Column
Efficient fractionating column designed with an an accurately fitted Dufton glass spiral. Quickfit™ Borosilicate Glass Dufton Fractionating Column can be disassembled for cleaning.
142.00 EUR - 805.00 EUR
Specifications
Type | Dufton Fractionating Column |
---|---|
Material | Borosilicate Glass |
Certifications/Compliance | ISO 3585 |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12001652
|
Quickfit™
FC1/12 |
270mm |
142.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12408906
|
Quickfit™
FC1/22 |
425mm |
157.00 EUR
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15507820
|
Quickfit™
FC2/13 |
290mm | N/A | ||||||
15517820
|
Quickfit™
FC2/23 |
300mm | N/A | ||||||
Description
- Glass column with accurately fitted Dufton glass spiral
- Good efficiency - liquid spiralling down column meets ascending vapour
- Spiral can be removed for cleaning
Specifications
Dufton Fractionating Column | |
Borosilicate Glass | |
ISO 3585 |