Learn More
Quickfit™ Borosilicate Glass Liebig Condenser
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
Brand:  Quickfit C1/44/SC
67.55 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12059652
  • 71.10 EUR / Each
Estimated Shipment: 11-07-2025
Description
Popular water-jacketed condenser suitable for distillation and reflux applications. The Quickfit Corosilicate Glass Liebig Condenser hase a detachable plastic screwthread connectors for hosing.
  • Complies with DIN 12576
  • Popular water-jacketed condenser suitable for distillation and reflux applications
  • With detachable plastic screwthread connectors for easy and safe attachment of rubber hose
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DIN 12576
550 mm
Borosilicate Glass
Suitable for distillation and reflux applications
29/32
Liebig Condenser
Documents
Product Information
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    1
    Hours
    21
    Minutes
    54
    Seconds
    45
  • Multiple Manufacturers
    Stock up on lab beakers, flasks and more before these deals disappear.
    Promo Code24123
    Expires2025-06-14
    New Offer
    Save Now
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Liebig Condenser > Cone Size: 29/32; Length: 550mm; Effective Length: 400mm