Learn More
Quickfit™ Expansion Socket Adapter
Made of borosilicate glass. Connects large sized ground glass joints with smaller sized ones. With the QuickFit™ expansion socket adaptor.
Brand:  Quickfit XA52
74.53 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 10427820
  • 78.45 EUR / Pack of 2
Estimated Shipment: 16-07-2025
Description
Connect large sized ground glass joints with smaller sized ones with the QuickFit™ expansion socket adaptor
  • Useful for the connection of dissimilar sized ground glass joints
  • Large sized socket at the top and small sized cone at the base
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19/26
Borosilicate Glass
34/35
Useful for the connection of dissimilar sized ground Glass joints
Expansion adapter
Documents
Product Information
Product Title
Quickfit™ Expansion Socket Adapter > Cone size: 19/26; Socket size: 34/35