Learn More
Quickfit™ Flat Flange Joint
Ideal for use as pipeline joints or in the construction of flat flange lids for use with wide neck reaction flasks.
Brand: Quickfit FG75
This item is not returnable.
View return policy
Description
- Highest quality flat flange joints
- Precision ground for leak free performance
- Choice of sizes up to 100mm bore
Specifications
0.17in. | |
4.4mm |
Product Content Correction
Product Title
Quickfit™ Flat Flange Joint > Outer Shank Dia.: 86.8mm; Outer Flange Dia.: 120mm; Bore size: 100mm; Thickness: 4.4mm
Spot an opportunity for improvement?Share a Content Correction