Learn More
Quickfit™ Recovery Still Head Adapter
Recover vapor after distillation. QuickFit™ glass recovery head provides an enclosed conduit between flasks and condensers.
Brand: Quickfit SH1/33
This item is not returnable.
View return policy
Description
Recover vapor after distillation. QuickFit™glass recovery head provides an enclosed conduit between flasks and condensers.
Provides a simple vapour conduit between flasks and distillation condensersSpecifications
Borosilicate Glass | |
24/29 (Flask/condenser) | |
Recovery Still Head Adapter |