Learn More
Quickfit™ Recovery Still Head Adapter
Recover vapor after distillation. QuickFit™ glass recovery head provides an enclosed conduit between flasks and condensers.
31.00 EUR - 51.65 EUR
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
11362105
|
Quickfit
SH1/11 |
14/23 (Flask/condenser) |
31.00 EUR
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
|||||
11372105
|
Quickfit
SH1/22 |
19/26 (Flask/condenser) |
36.55 EUR
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
|||||
11382105
|
Quickfit
SH1/23 |
24/29 (Flask), 19/26 (condenser) |
38.55 EUR
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
|||||
11392105
|
Quickfit
SH1/33 |
24/29 (Flask/condenser) |
42.40 EUR
Each |
Estimated Shipment: 05-05-2025
Log in to see stock. |
|||||
11302115
|
Quickfit
SH1/44 |
29/32 (Flask/condenser) |
51.65 EUR
Each |
Estimated Shipment: 25-04-2025
Log in to see stock. |
|||||
Description
Recover vapor after distillation. QuickFit™glass recovery head provides an enclosed conduit between flasks and condensers.
Provides a simple vapour conduit between flasks and distillation condensersSpecifications
Borosilicate Glass | |
Recovery Still Head Adapter |