Learn More
(R)-1-(3-Chlorophenyl)ethylamine, ChiPros™, 99%, ee 98+%, Thermo Scientific Chemicals
101.00 EUR - 305.52 EUR
Chemical Identifiers
CAS | 17061-53-9 |
---|---|
Molecular Formula | C8H10ClN |
Molecular Weight (g/mol) | 155.625 |
MDL Number | MFCD06761822 |
InChI Key | DQEYVZASLGNODG-ZCFIWIBFSA-N |
Synonym | r-1-3-chlorophenyl ethanamine, 1r-1-3-chlorophenyl ethanamine, r-1-3-chlorophenyl ethylamine, r-3-chloro-, a-methylbenzylamine, 1r-1-3-chlorophenyl ethan-1-amine, r-3-chloro-methylbenzylamine, r-+-1-3-chlorophenyl ethylamine, r-1-3-chlorophenyl ethylamine, chipros, r-alpha-methyl-3-chlorobenzenemethaneamine, benzenemethanamine,3-chloro-a-methyl-, ar |
PubChem CID | 2507697 |
IUPAC Name | (1R)-1-(3-chlorophenyl)ethanamine |
SMILES | CC(C1=CC(=CC=C1)Cl)N |
Description
(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-1-(3-Chlorophenyl)ethylamine play a very extensive role of making chirally optically active drugs and compounds. Also as intermediates.
Solubility
Insoluble in water.
Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
17061-53-9 | |
155.625 | |
DQEYVZASLGNODG-ZCFIWIBFSA-N | |
2507697 | |
CC(C1=CC(=CC=C1)Cl)N |
C8H10ClN | |
MFCD06761822 | |
r-1-3-chlorophenyl ethanamine, 1r-1-3-chlorophenyl ethanamine, r-1-3-chlorophenyl ethylamine, r-3-chloro-, a-methylbenzylamine, 1r-1-3-chlorophenyl ethan-1-amine, r-3-chloro-methylbenzylamine, r-+-1-3-chlorophenyl ethylamine, r-1-3-chlorophenyl ethylamine, chipros, r-alpha-methyl-3-chlorobenzenemethaneamine, benzenemethanamine,3-chloro-a-methyl-, ar | |
(1R)-1-(3-chlorophenyl)ethanamine |
Specifications
17061-53-9 | |
C8H10ClN | |
UN2735 | |
Insoluble in water. | |
CC(C1=CC(=CC=C1)Cl)N | |
155.625 | |
155.63 | |
ee ≥98% |
112°C (13 mmHg) | |
MFCD06761822 | |
r-1-3-chlorophenyl ethanamine, 1r-1-3-chlorophenyl ethanamine, r-1-3-chlorophenyl ethylamine, r-3-chloro-, a-methylbenzylamine, 1r-1-3-chlorophenyl ethan-1-amine, r-3-chloro-methylbenzylamine, r-+-1-3-chlorophenyl ethylamine, r-1-3-chlorophenyl ethylamine, chipros, r-alpha-methyl-3-chlorobenzenemethaneamine, benzenemethanamine,3-chloro-a-methyl-, ar | |
DQEYVZASLGNODG-ZCFIWIBFSA-N | |
(1R)-1-(3-chlorophenyl)ethanamine | |
2507697 | |
99% | |
(R)-1-(3-Chlorophenyl)ethylamine |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures