missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-Amino-3-(4-cyanophenyl)propionic acid, 95%, Thermo Scientific™ Product Code.: 15469615

(R)-3-Amino-3-(4-cyanophenyl)propionic acid, 95%, Thermo Scientific™

Product Code. 15469615
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15469615

Brand: Thermo Scientific Alfa Aesar H52132.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 738606-24-1
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD04113665
InChI Key JFPLLJRLBHIJPS-SECBINFHSA-N
Synonym r-3-amino-3-4-cyanophenyl propanoic acid, r-3-amino-3-4-cyano-phenyl-propionic acid, 3r-3-amino-3-4-cyanophenyl propanoic acid, r-3-amino-3-4-cyanophenyl propionic acid, h-d-b-phe 4-cn-oh, r-beta-amino-4-cyanohydrocinnamic acid, r-3-amino-3-4-cyanophenyl propanoicacid, benzenepropanoic acid, b-amino-4-cyano-, br
PubChem CID 1491713
IUPAC Name (3R)-3-amino-3-(4-cyanophenyl)propanoic acid
SMILES C1=CC(=CC=C1C#N)C(CC(=O)O)N

Specifications

Quantity 250 mg
Formula Weight 190.2
Sensitivity Air Sensitive
Percent Purity 95%
Chemical Name or Material (R)-3-Amino-3-(4-cyanophenyl)propionic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK02W018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberH14Y023 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-Amino-3-(4-cyanophenyl)propionic acid, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.