missing translation for 'onlineSavingsMsg'
Learn More
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid, 95%, Thermo Scientific Chemicals Product Code.: 15409645

(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid, 95%, Thermo Scientific Chemicals

Product Code. 15409645
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15409645

Brand: Thermo Scientific Alfa Aesar H52166.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261380-20-5
Molecular Formula C14H18BrNO4
Molecular Weight (g/mol) 344.205
MDL Number MFCD01320860
InChI Key ZAMLGGRVTAXBHI-LLVKDONJSA-N
Synonym boc-r-3-amino-3-4-bromo-phenyl-propionic acid, r-3-4-bromophenyl-3-tert-butoxycarbonyl amino propanoic acid, r-boc-4-bromo-beta-phe-oh, boc-4-bromo-l-beta-phenylalanine, boc-beta-r-4-bromophenylalanine, r-n-boc-3-amino-3-4-bromophenyl propanoic acid, boc-r-4-beta-bromophenylalanine, r-3-boc-amino-3-4-bromophenyl propionic acid, boc-r-4-bromo-beta-phenylalanine, r-n-boc-beta-4-bromophenyl-beta-alanine
PubChem CID 7009110
IUPAC Name (3R)-3-(4-bromophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=C(C=C1)Br

Specifications

Melting Point 143°C to 145°C
Quantity 250 mg
Formula Weight 344.2
Percent Purity 95%
Chemical Name or Material (R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS07E028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52166.03,H52166.06,H52166.MD
Lot NumberG18X046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52166.03,H52166.06,H52166.MD
Lot NumberK19W098 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52166.03,H52166.06,H52166.MD
Lot NumberY02D010 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH52166.03,H52166.06,H52166.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-3-(Boc-amino)-3-(4-bromophenyl)propionic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.