Learn More
Fisherbrand™ Raised Shelves for Stainless Steel Tanks
Raised shelves for use with stainless steel tanks.
116.00 EUR - 229.00 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
17571906
|
Fisherbrand™
17571906 |
Fisherbrand™ Digital Heated Circulating Bath, 12 L |
116.00 EUR
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
17581906
|
Fisherbrand™
17581906 |
Fisherbrand™ Digital Heated Circulating Bath, 18 L |
169.00 EUR
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
17591906
|
Fisherbrand™
17591906 |
Fisherbrand™ Digital Heated Circulating Bath, 26 L |
179.00 EUR
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
17501916
|
Fisherbrand™
17501916 |
Fisherbrand™ Digital Heated Circulating Bath, 38 L |
229.00 EUR
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- For use with digital heated circulating baths
- Reversible
- Allows for two shelf depths
Specifications
Raised Shelf |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Raised Shelves for Stainless Steel Tanks