missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant H. pylori Glutamate--tRNA ligase 1 Protein
A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Glutamate--tRNA ligase 1, was expressed in the hosts and tags indicated. option, abo
Brand: enQuireBio™ QP6981-ye-1mg
This item is not returnable.
View return policy
Specifications
NP_207274.1 | |
Glutamate--tRNA ligase 1 Protein | |
Research Use Only | |
gltX1 | |
Recombinant Protein | |
Yeast | |
MSLIVTRFAP SPTGYLHIGG LRTAIFNYLF ARANQGKFFL RIEDTDLSRN SIEAANAIIE AFKWVGLEYD GEILYQSKRF EIYKEYIQKL LDEDKAYYCY MSKEELDALR EEQKARKETP RYDNRYRDFK GTPPKGIEPV VRIKVPQNEV IGFNDGVKGE VKVNTNELDD FIIARSDGTP TYNFVVTIDD ALMGITDVIR GDDHLSNTPK QIVLYKALNF KIPNFFHVPM ILNEEGQKLS KRHGATNVMD YQEMGYLKEA LVNFLARLGW SYQDKEVFSM QELLELFDPK DLNSSPSCFS WHKLNWLNAH YLKNQSVQEL LKLLKPFSFS DLSHLNPTQL DRLLDALKER SQTLKELALK IDEVLIAPVE YEEKVFKKLN QALVMPLLEK FKLELNKANF NDESALENAM RQIIEEEKIK AGSFMQPLRL ALLGKGGGIG LKEALFILGK TESVKRIEDF LKN | |
Greater than 90% as determined by SDS-PAGE. |
898901 | |
1 mg | |
Not determined. | |
Full Length | |
H. pylori | |
His | |
Tris-based buffer, 50% glycerol |