missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant H. pylori Glutamate--tRNA ligase 1 Protein

A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) Glutamate--tRNA ligase 1, was expressed in the hosts and tags indicated. option, abo

352.00 EUR - 3175.00 EUR

Specifications

Accession Number NP_207274.1
Gene ID (Entrez) 898901
Name Glutamate--tRNA ligase 1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16096920
View Documents Promotion Details
enQuireBio™
QP6981-ye-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16036930
View Documents Promotion Details
enQuireBio™
QP6981-ye-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16086920
View Documents Promotion Details
enQuireBio™
QP6981-ye-100ug
100 μg
1145.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
16016930
View Documents Promotion Details
enQuireBio™
QP6981-ye-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16026930
View Documents Promotion Details
enQuireBio™
QP6981-ye-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16006930
View Documents Promotion Details
enQuireBio™
QP6981-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_207274.1
Glutamate--tRNA ligase 1 Protein
Not determined.
Full Length
H. pylori
His
Tris-based buffer, 50% glycerol
898901
Research Use Only
gltX1
Recombinant Protein
Yeast
MSLIVTRFAP SPTGYLHIGG LRTAIFNYLF ARANQGKFFL RIEDTDLSRN SIEAANAIIE AFKWVGLEYD GEILYQSKRF EIYKEYIQKL LDEDKAYYCY MSKEELDALR EEQKARKETP RYDNRYRDFK GTPPKGIEPV VRIKVPQNEV IGFNDGVKGE VKVNTNELDD FIIARSDGTP TYNFVVTIDD ALMGITDVIR GDDHLSNTPK QIVLYKALNF KIPNFFHVPM ILNEEGQKLS KRHGATNVMD YQEMGYLKEA LVNFLARLGW SYQDKEVFSM QELLELFDPK DLNSSPSCFS WHKLNWLNAH YLKNQSVQEL LKLLKPFSFS DLSHLNPTQL DRLLDALKER SQTLKELALK IDEVLIAPVE YEEKVFKKLN QALVMPLLEK FKLELNKANF NDESALENAM RQIIEEEKIK AGSFMQPLRL ALLGKGGGIG LKEALFILGK TESVKRIEDF LKN
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documentos

Documentos

Certificados