missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HPV HPV 6 Protein

A cDNA sequence encoding the HPV 6 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12309-1mg

 View more versions of this product

Product Code. 15960009

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

HPV HPV 6 Protein
Research Use Only
Recombinant Protein
E. coli
VDVPPPNPVSKVVATDAYVTRTNIFYHASSSRLLAVGHPYFSIKRANKTVVPKVSGYQYRVFKVVLPDPNKFALPDSSLFDPTTQRLVWACTGLEVGRGQPLGVGVSGHPFLNKYDDVENSGSGGNPGQDNRVNVGMDYKQTQLCMVGCAPPLGEHWGKGKQCTNTPVQAGDCPPLELITSVIQDGDMVDTGFGAMNFADLQTNKSDVPIDICGTTCKYPDYLQMAADPYGDRLFFFLRKEQMFARHFFNRAGEVGEPVPDTLIIKGSGNRTSVGSSIYVNTPSGSLVSSEAQLFNKPYWLQKAQGHNNGICWGNQLFVTVVDTTRSTNMTLCASVTTSSTYTNSDYKEYMRHVEEYDLQFIFQLCSITLSAEVMAYIHTMNPSVLEDWNFGLSPPPNGTLEDTYRYVQSQAITCQKPTPEKEKPDPYKNLSFWEVNLKEKFSSELDQYPLGRKFLLQSGYRGRSSIRTGVKRPAVSKASAAPKRKRAK
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HPV
GST
PBS and 3M Urea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers