missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HPV HPV 6 Protein
A cDNA sequence encoding the HPV 6 was constructed and used to recombinantly synthesize the protein.
Specifications
Name | HPV HPV 6 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HPV |
Specifications
HPV HPV 6 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HPV | |
GST | |
PBS and 3M Urea. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
VDVPPPNPVSKVVATDAYVTRTNIFYHASSSRLLAVGHPYFSIKRANKTVVPKVSGYQYRVFKVVLPDPNKFALPDSSLFDPTTQRLVWACTGLEVGRGQPLGVGVSGHPFLNKYDDVENSGSGGNPGQDNRVNVGMDYKQTQLCMVGCAPPLGEHWGKGKQCTNTPVQAGDCPPLELITSVIQDGDMVDTGFGAMNFADLQTNKSDVPIDICGTTCKYPDYLQMAADPYGDRLFFFLRKEQMFARHFFNRAGEVGEPVPDTLIIKGSGNRTSVGSSIYVNTPSGSLVSSEAQLFNKPYWLQKAQGHNNGICWGNQLFVTVVDTTRSTNMTLCASVTTSSTYTNSDYKEYMRHVEEYDLQFIFQLCSITLSAEVMAYIHTMNPSVLEDWNFGLSPPPNGTLEDTYRYVQSQAITCQKPTPEKEKPDPYKNLSFWEVNLKEKFSSELDQYPLGRKFLLQSGYRGRSSIRTGVKRPAVSKASAAPKRKRAK | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |