missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARHI / DIRAS3 Protein
A DNA sequence encoding the Homo sapiens (Human) ARHI / DIRAS3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5935-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_004666.1 | |
ARHI / DIRAS3 Protein | |
Research Use Only | |
DIRAS3 | |
Recombinant Protein | |
E. coli | |
MGNASFGSKE QKLLKRLRLL PALLILRAFK PHRKIRDYRV VVVGTAGVGK STLLHKWASG NFRHEYLPTI ENTYCQLLGC SHGVLSLHIT DSKSGDGNRA LQRHVIARGH AFVLVYSVTK KETLEELKAF YELICKIKGN NLHKFPIVLV GNKSDDTHRE VALNDGATCA MEWNCAFMEI SAKTDVNVQE LFHMLLNYKK KPTTGLQEPE KKSQMPNTTE KLLDKC | |
Greater than 80% as determined by SDS-PAGE. |
9077 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |