missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ARHI / DIRAS3 Protein

A DNA sequence encoding the Homo sapiens (Human) ARHI / DIRAS3, was expressed in the hosts and tags indicated.

245.00 EUR - 2985.00 EUR

Specifications

Accession Number NP_004666.1
Gene ID (Entrez) 9077
Name ARHI / DIRAS3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15948779
View Documents Promotion Details
enQuireBio™
QP5935-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15988779
View Documents Promotion Details
enQuireBio™
QP5935-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15938779
View Documents Promotion Details
enQuireBio™
QP5935-ec-100ug
100 μg
675.00 EUR
100µg
Please to purchase this item. Need a web account? Register with us today!
15968779
View Documents Promotion Details
enQuireBio™
QP5935-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15978779
View Documents Promotion Details
enQuireBio™
QP5935-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15958779
View Documents Promotion Details
enQuireBio™
QP5935-ec-1mg
1 mg
2985.00 EUR
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_004666.1
ARHI / DIRAS3 Protein
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
9077
Research Use Only
DIRAS3
Recombinant Protein
E. coli
MGNASFGSKE QKLLKRLRLL PALLILRAFK PHRKIRDYRV VVVGTAGVGK STLLHKWASG NFRHEYLPTI ENTYCQLLGC SHGVLSLHIT DSKSGDGNRA LQRHVIARGH AFVLVYSVTK KETLEELKAF YELICKIKGN NLHKFPIVLV GNKSDDTHRE VALNDGATCA MEWNCAFMEI SAKTDVNVQE LFHMLLNYKK KPTTGLQEPE KKSQMPNTTE KLLDKC
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications