missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EMCN Protein
A cDNA sequence encoding the EMCN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11782-2ug
This item is not returnable.
View return policy
Specifications
51705 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMNSTGV LEAANNSLVV TTTKPSITTP NTESLQKNVV TPTTGTTPKG TITNELLKMS LMSTATFLTS KDEGLKATTT DVRKNDSIIS NVTVTSVTLP NAVSTLQSSK PKTETQSSIK TTEIPGSVLQ PDASPSKTGT LTSIPVTIPE NTSQSQVIGT EGGKNASTSA TSRSYSS | |
Greater than 90% as determined by SDS-PAGE. |
EMCN Protein | |
Research Use Only | |
EMCN | |
Human | |
His | |
EMCN protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 100mM NaCl. |