missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EMCN Protein
A cDNA sequence encoding the EMCN was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 1595.00 EUR
Specifications
Gene ID (Entrez) | 51705 |
---|---|
Name | EMCN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | EMCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958348
|
enQuireBio™
QP11782-2ug |
2 μg |
120.00 EUR
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15948348
|
enQuireBio™
QP11782-10ug |
10 μg |
186.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15938348
|
enQuireBio™
QP11782-100ug |
100 μg |
1595.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
51705 | |
Research Use Only | |
EMCN | |
Human | |
His | |
EMCN protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 10% glycerol, 1mM DTT and 100mM NaCl. |
EMCN Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMNSTGV LEAANNSLVV TTTKPSITTP NTESLQKNVV TPTTGTTPKG TITNELLKMS LMSTATFLTS KDEGLKATTT DVRKNDSIIS NVTVTSVTLP NAVSTLQSSK PKTETQSSIK TTEIPGSVLQ PDASPSKTGT LTSIPVTIPE NTSQSQVIGT EGGKNASTSA TSRSYSS | |
Greater than 90% as determined by SDS-PAGE. |