missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGA Protein
A DNA sequence encoding the Homo sapiens (Human) FGA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6034-ec-1mg
This item is not returnable.
View return policy
Specifications
FGA Protein | |
Research Use Only | |
FGA | |
Recombinant Protein | |
E. coli | |
GPRVVERHQS ACKDSDWPFC SGEDWNYKCP SGCRMKGLID EVNQDFTNRI NKLKNSLFEY QKNNKDSHSL TTNIMEILRG DFSSANNRDN TYNRVSEDLR SRIEVLKRKV IEKVTANNLL VARVTTEETP HLKARAIKWQ MRPEVKPIMK EHIAPREAML NLALSEVSTL LLWGSLPCPP RLS | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |