missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGA Protein
A DNA sequence encoding the Homo sapiens (Human) FGA, was expressed in the hosts and tags indicated.
186.00 EUR - 1855.00 EUR
Specifications
Name | FGA Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | FGA |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969509
|
enQuireBio™
QP6034-ec-10ug |
10 μg |
186.00 EUR
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15909519
|
enQuireBio™
QP6034-ec-50ug |
50 μg | N/A | ||||||
15959509
|
enQuireBio™
QP6034-ec-100ug |
100 μg |
463.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15989509
|
enQuireBio™
QP6034-ec-200ug |
200 μg | N/A | ||||||
15999509
|
enQuireBio™
QP6034-ec-500ug |
500 μg |
1330.00 EUR
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15979509
|
enQuireBio™
QP6034-ec-1mg |
1 mg | N/A | ||||||
Specifications
FGA Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
FGA | |
Recombinant Protein | |
E. coli | |
GPRVVERHQS ACKDSDWPFC SGEDWNYKCP SGCRMKGLID EVNQDFTNRI NKLKNSLFEY QKNNKDSHSL TTNIMEILRG DFSSANNRDN TYNRVSEDLR SRIEVLKRKV IEKVTANNLL VARVTTEETP HLKARAIKWQ MRPEVKPIMK EHIAPREAML NLALSEVSTL LLWGSLPCPP RLS | |
Greater than 80% as determined by SDS-PAGE. |