missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS23 Protein
A cDNA sequence encoding the MRPS23 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12733-1mg
This item is not returnable.
View return policy
Specifications
MRPS23 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGSRLE TVGSIFSRTR DLVRAGVLKE KPLWFDVYDA FPPLREPVFQ RPRVRYGKAK APIQDIWYHE DRIRAKFYSV YGSGQRAFDL FNPNFKSTCQ RFVEKYTELQ KLGETDEEKL FVETGKALLA EGVILRRVGE ARTQHGGSHV SRKSEHLSVR PQTALEENET QKEVPQDQHL EAPADQSKGL LPP | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPS23 protein solution (1 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |