missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MRPS23 Protein
A cDNA sequence encoding the MRPS23 was constructed and used to recombinantly synthesize the protein.
120.00 EUR - 3585.00 EUR
Specifications
Name | MRPS23 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961369
|
enQuireBio™
QP12733-5ug |
5 μg |
120.00 EUR
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15951369
|
enQuireBio™
QP12733-20ug |
20 μg |
186.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15941369
|
enQuireBio™
QP12733-1mg |
1 mg |
3585.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
MRPS23 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MRPS23 protein solution (1 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGSRLE TVGSIFSRTR DLVRAGVLKE KPLWFDVYDA FPPLREPVFQ RPRVRYGKAK APIQDIWYHE DRIRAKFYSV YGSGQRAFDL FNPNFKSTCQ RFVEKYTELQ KLGETDEEKL FVETGKALLA EGVILRRVGE ARTQHGGSHV SRKSEHLSVR PQTALEENET QKEVPQDQHL EAPADQSKGL LPP | |
Greater than 85% as determined by SDS-PAGE. |