missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Cathepsin S / CTSS Protein

A DNA sequence encoding the Mus musculus (Mouse) Cathepsin S / CTSS, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5895-ec-500ug

 View more versions of this product

Product Code. 15978509

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_067256.4
Cathepsin S / CTSS Protein
Research Use Only
ctss
Recombinant Protein
E. coli
LPDTVDWREK GCVTEVKYQG SCGACWAFSA VGALEGQLKL KTGKLISLSA QNLVDCSNEE KYGNKGCGGG YMTEAFQYII DNGGIEADAS YPYKATDEKC HYNSKNRAAT CSRYIQLPFG DEDALKEAVA TKGPVSVGID ASHSSFFFYK SGVYDDPSCT GNVNHGVLVV GYGTLDGKDY WLVKNSWGLN FGDQGYIRMA RNNKNHCGIA SYCSYPEI
Greater than 90% as determined by SDS-PAGE.
13040
500 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers