missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cathepsin S / CTSS Protein
A DNA sequence encoding the Mus musculus (Mouse) Cathepsin S / CTSS, was expressed in the hosts and tags indicated.
245.00 EUR - 2985.00 EUR
Specifications
Accession Number | NP_067256.4 |
---|---|
Gene ID (Entrez) | 13040 |
Name | Cathepsin S / CTSS Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948509
|
enQuireBio™
QP5895-ec-10ug |
10 μg | N/A | ||||||
15988509
|
enQuireBio™
QP5895-ec-50ug |
50 μg | N/A | ||||||
15938509
|
enQuireBio™
QP5895-ec-100ug |
100 μg |
675.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15968509
|
enQuireBio™
QP5895-ec-200ug |
200 μg | N/A | ||||||
15978509
|
enQuireBio™
QP5895-ec-500ug |
500 μg | N/A | ||||||
15958509
|
enQuireBio™
QP5895-ec-1mg |
1 mg |
2985.00 EUR
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_067256.4 | |
Cathepsin S / CTSS Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
13040 | |
Research Use Only | |
ctss | |
Recombinant Protein | |
E. coli | |
LPDTVDWREK GCVTEVKYQG SCGACWAFSA VGALEGQLKL KTGKLISLSA QNLVDCSNEE KYGNKGCGGG YMTEAFQYII DNGGIEADAS YPYKATDEKC HYNSKNRAAT CSRYIQLPFG DEDALKEAVA TKGPVSVGID ASHSSFFFYK SGVYDDPSCT GNVNHGVLVV GYGTLDGKDY WLVKNSWGLN FGDQGYIRMA RNNKNHCGIA SYCSYPEI | |
Greater than 90% as determined by SDS-PAGE. |