missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cathepsin S / CTSS Protein
A DNA sequence encoding the Mus musculus (Mouse) Cathepsin S / CTSS, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5895-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_067256.4 | |
Cathepsin S / CTSS Protein | |
Research Use Only | |
ctss | |
Recombinant Protein | |
E. coli | |
LPDTVDWREK GCVTEVKYQG SCGACWAFSA VGALEGQLKL KTGKLISLSA QNLVDCSNEE KYGNKGCGGG YMTEAFQYII DNGGIEADAS YPYKATDEKC HYNSKNRAAT CSRYIQLPFG DEDALKEAVA TKGPVSVGID ASHSSFFFYK SGVYDDPSCT GNVNHGVLVV GYGTLDGKDY WLVKNSWGLN FGDQGYIRMA RNNKNHCGIA SYCSYPEI | |
Greater than 90% as determined by SDS-PAGE. |
13040 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |