Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Brand:  BINDER™ 8012-2525
 View more versions of this product
Product Code. 30238272
  • 1032.00 EUR / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Description
  • Reinforced inner chamber for bulky and heavy samples.
  • Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
Specifications
KBF PRO 260
Stainless Steel
Chamber Rack
180 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Reinforced Inner Chamber > For use with KBF PRO 260