Learn More
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Mounting Fluid
Remel PathoDx and Respiratory Virus Panel Mounting Fluid is used with PathoDx Respiratory Virus Panel.
Brand:  Thermo Scientific™ R62413
Product Code. 11404139
Description
Description
Remel PathoDx and Respiratory Virus Panel Mounting Fluid is used with PathoDx Respiratory Virus Panel.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PathoDx™ Respiratory Virus Panel Respiratory Mounting Fluid
Mounting Fluid
1 x 15 mL
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3491407 Certificate TypeCertificate of Analysis Date27/05/2022 Catalog NumberR62413
Lot Number3476138 Certificate TypeCertificate of Analysis Date26/04/2022 Catalog NumberR62413
Lot Number3445464 Certificate TypeCertificate of Analysis Date11/03/2022 Catalog NumberR62413
Lot Number3354705 Certificate TypeCertificate of Analysis Date09/10/2021 Catalog NumberR62413
Lot Number3297079 Certificate TypeCertificate of Analysis Date21/05/2021 Catalog NumberR62413
Product Title
Thermo Scientific™ Remel™ PathoDX™ Respiratory Virus Panel Mounting Fluid > PathoDx™ Respiratory Virus Panel Respiratory Mounting Fluid