Learn More
Grant Instruments™ Polycarbonate Transparent Lid, Gabled
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ AQL26
152.35 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
 View more versions of this product
Product Code. 11406427
  • 162.00 EUR / Each
Estimated Shipment: 28-05-2025
Description
Directs condensation away from immersed vessels, avoids contamination, reduces evaporation and saves energy.
  • Polycarbonate construction
  • Clear, gabled lid allows condensation buildup to drip away from immersed vessels.
  • For use with Sub Aqua Pro Series Unstirred Water Baths
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gabled
26L Tanks
Polycarbonate
Product Title
Grant Instruments™ Polycarbonate Transparent Lid, Gabled > For 26L Tanks